Acetanilide, Sigma-Aldrich®

Supplier: Merck
Warning


M-Clarity™ quality level = MQ200

112933-100G 112933-1KG 112933-500G 112933-5G
SIAL112933-100GEA 40.4 EUR
SIAL112933-100G SIAL112933-1KG SIAL112933-500G SIAL112933-5G
Acetanilide, Sigma-Aldrich®
Acetanilide
Formula: C₈H₉NO
Boiling Pt: 304 °C (1013 hPa)
Melting Pt: 115 °C
Density: 1.22 g/cm³ (20 °C)
Flash Pt: 174 °C
MDL Number: MFCD00008674
CAS Number: 103-84-4
EINECS: 203-150-7
Merck Index: 13,00051

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR