Acetanilide, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ100
397229-25G 397229-5G
SIAL397229-25GEA 288 EUR
SIAL397229-25G SIAL397229-5G
Acetanilide, Sigma-Aldrich®
Acetanilide
Formula: C₈H₉NO
Boiling Pt: 304 °C (1013 hPa)
Melting Pt: 115 °C
Density: 1.22 g/cm³ (20 °C)
Flash Pt: 174 °C
MDL Number: MFCD00008674
CAS Number: 103-84-4
EINECS: 203-150-7
Merck Index: 13,00051