Metacetamol, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: N-(3-Hydroxyphenyl)acetamide, N-Acetyl-3-aminophenol, Paracetamol, 3-Acetamidophenol, 3'-Hydroxyacetanilide
M-Clarity™ quality level = MQ200
A7205-100G A7205-25G
SIALA7205-100GEA 195 EUR
SIALA7205-100G SIALA7205-25G
Metacetamol, Sigma-Aldrich®
Metacetamol
Formula: C₈H₉NO₂
MW: 151.16 g/mol
Boiling Pt: 351 °C (1013 hPa)
Melting Pt: 147…150 °C
Storage Temperature: Ambient
MDL Number: MFCD00002263
CAS Number: 621-42-1
EINECS: 210-687-0