Ethyl butyrate, Sigma-Aldrich®
Supplier: Merck
Synonyms:
Butanoic acid ethyl ester, Butyric acid ethyl ester
M-Clarity™ quality level = MQ300
W242713-1KG-K
W242713-20KG-K
W242713-4KG-K
W242713-9KG-K
SIALW242713-1KG-KEA
90.1
EUR
SIALW242713-1KG-K
SIALW242713-20KG-K
SIALW242713-4KG-K
SIALW242713-9KG-K
Ethyl butyrate, Sigma-Aldrich®
Ethyl butyrate
Formula:
C₆H₁₂O₂ MW: 116.16 g/mol Boiling Pt: 252 °C (1013 hPa) Melting Pt: –92 °C Density: 0.88 g/cm³ (20 °C) Flash Pt: 26 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009394 CAS Number: 105-54-4 EINECS: 203-306-4 UN: 1180 ADR: 3,III Merck Index: 13,03810 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...