Methyl butyrate, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ300
W269328-100G-K W269328-1KG-K W269328-4KG-K
SIALW269328-100G-KEA 89.7 EUR
SIALW269328-100G-K SIALW269328-1KG-K SIALW269328-4KG-K
Methyl butyrate, Sigma-Aldrich®
Methyl butyrate
Formula: C₅H₁₀O₂
Boiling Pt: 102 °C (1013 hPa)
Melting Pt: –84 °C
Density: 0.8984 g/cm³ (20 °C)
Flash Pt: 14 °C
MDL Number: MFCD00009391
CAS Number: 623-42-7
EINECS: 210-792-1
UN: 1237
ADR: 3,II
Merck Index: 13,06062