Isobutyl acetate
Supplier: EHRENSTORFER GMBH
EHERC10016100EA
47.2
EUR
EHERC10016100
Isobutyl acetate
Isobutyl acetate
Formula:
C₆H₁₂O₂ MW: 116,16 g/mol Boiling Pt: 118 °C (1013 hPa) Melting Pt: –99 °C Density: 0,87 g/cm³ (20 °C) Flash Pt: 18 °C |
MDL Number:
MFCD00008932 UN: 1213 ADR: 3,II Merck Index: 13,05147 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...