Isobutyl acetate, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ300

SIALW217514-4KG-KEA 341 EUR
SIALW217514-4KG-K SIALW217514-1KG-K
Isobutyl acetate, Sigma-Aldrich®
Isobutyl acetate
Formula: C₆H₁₂O₂
MW: 116,16 g/mol
Boiling Pt: 118 °C (1013 hPa)
Melting Pt: –99 °C
Density: 0,87 g/cm³ (20 °C)
Flash Pt: 18 °C
MDL Number: MFCD00008932
UN: 1213
ADR: 3,II
Merck Index: 13,05147

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR