tert-Butyl acetate, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: 1,1-Dimethylethyl acetate
M-Clarity™ quality level = MQ200
B88209-100ML B88209-500ML B88209-5ML
SIALB88209-100MLEA 73.1 EUR
SIALB88209-100ML SIALB88209-500ML SIALB88209-5ML
tert-Butyl acetate, Sigma-Aldrich®
tert-Butyl acetate
Formula: C₆H₁₂O₂
MW: 116.16 g/mol
Boiling Pt: 98
Melting Pt: 90…97 °C
Density: 0.87 g/cm³ (20 °C)
Flash Pt: 15
Storage Temperature: Ambient
MDL Number: MFCD00008807
CAS Number: 540-88-5
EINECS: 208-760-7
UN: 1123
ADR: 3 II
Merck Index: 13,01536