Isopropyl acetate, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Acetic acid isopropyl ester
M-Clarity™ quality level = MQ300
W292605-1KG-K W292605-20KG-K W292605-9KG-K
SIALW292605-1KG-KEA 82.9 EUR
SIALW292605-1KG-K SIALW292605-20KG-K SIALW292605-9KG-K
Isopropyl acetate, Sigma-Aldrich®
Isopropyl acetate
Formula: C₅H₁₀O₂
MW: 102.13 g/mol
Boiling Pt: 85 °C (1013 hPa)
Melting Pt: –62 °C
Density: 0.8732 g/cm³ (20 °C)
Flash Pt: 4 °C
Storage Temperature: Ambient
MDL Number: MFCD00008877
CAS Number: 108-21-4
EINECS: 203-561-1
UN: 1220
ADR: 3,II
Merck Index: 13,05225