Heptyl acetate, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ300
W254711-100G-K
W254711-1KG-K
SIALW254711-100G-KEA
159
EUR
SIALW254711-100G-K
SIALW254711-1KG-K
Heptyl acetate, Sigma-Aldrich®
Heptyl acetate
Formula:
C₉H₁₈O₂ Boiling Pt: 190 °C (1013 hPa) Melting Pt: –50.2 °C Density: 0.8891 g/cm³ (0 °C) |
MDL Number:
MFCD00027311 CAS Number: 112-06-1 EINECS: 203-932-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...