Methyl acetate, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Acetic acid methyl ester
M-Clarity™ quality level = MQ300
W267600-10KG W267600-1KG W267600-20KG W267600-5KG W267600-SAMPLE
SIALW267600-10KGEA 234 EUR
SIALW267600-10KG SIALW267600-1KG SIALW267600-20KG SIALW267600-5KG SIALW267600-SAMPLE
Methyl acetate, Sigma-Aldrich®
Methyl acetate
Formula: C₃H₆O₂
MW: 74.08 g/mol
Boiling Pt: 56.9 °C (1013 hPa)
Melting Pt: –98 °C
Density: 0.938 g/cm³ (20 °C)
Flash Pt: –13 °C
Storage Temperature: Ambient
MDL Number: MFCD00008711
CAS Number: 79-20-9
EINECS: 201-185-2
UN: 1231
ADR: 3,II
Merck Index: 13,06038