Octyl acetate, Sigma-Aldrich®

Supplier: Merck

Synonyms: Acetic acid octyl ester

M-Clarity™ quality level = MQ300

W280615-100G W280615-1KG W280615-25G W280615-SAMPLE
SIALW280615-100GEA 128 EUR
SIALW280615-100G SIALW280615-1KG SIALW280615-25G SIALW280615-SAMPLE
Octyl acetate, Sigma-Aldrich®
Octyl acetate
Formula: C₁₀H₂₀O₂
MW: 172.27 g/mol
Boiling Pt: 195…199 °C (1013 hPa)
Melting Pt: –38.5 °C
Density: 0.885 g/cm³ (0 °C)
MDL Number: MFCD00009562
CAS Number: 112-14-1
EINECS: 203-939-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR