Amyl acetate, Sigma-Aldrich®
Supplier: Merck
Danger
109584-1L 109584-250ML
SIAL109584-1LEA 169 EUR
SIAL109584-1L SIAL109584-250ML
Amyl acetate, Sigma-Aldrich®
Amyl acetate
Formula: C₇H₁₄O₂
Boiling Pt: 149 °C (1013 hPa)
Melting Pt: –71 °C
Density: 0.876 g/cm³ (20 °C)
Flash Pt: 23 °C
Storage Temperature: Ambient
MDL Number: MFCD00009500
CAS Number: 628-63-7
EINECS: 211-047-3
UN: 1104
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=37af9fce9b4d1452153c5c88d6e47d0d5e08f221&vid=b0e4dd5050dab83dda2db704a160c2058b4bd15a&prid=web&clid=SAPDAM