Amyl acetate, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ300
SIALW504009-4KGEA 247 EUR
SIALW504009-4KG SIALW504009-500G SIALW504009-1KG SIALW504009-SAMPLE SIALW504009-20KG
Amyl acetate, Sigma-Aldrich®
Amyl acetate
Formula: C₇H₁₄O₂
MW: 130,19 g/mol
Boiling Pt: 149 °C (1013 hPa)
Melting Pt: –71 °C
Density: 0,876 g/cm³ (20 °C)
Flash Pt: 23 °C
MDL Number: MFCD00009500
UN: 1104
ADR: 3,III
https://si.vwr.com/stibo/web/std.lang.all/27/75/722775.jpg