Propyl acetate, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Acetic acid propyl ester
M-Clarity™ quality level = MQ100
W292516-100G-K W292516-1KG-K
SIALW292516-100G-KEA 95.7 EUR
SIALW292516-100G-K SIALW292516-1KG-K
Propyl acetate, Sigma-Aldrich®
Propyl acetate
Formula: C₅H₁₀O₂
MW: 102.13 g/mol
Boiling Pt: 102
Melting Pt: –95 °C
Density: 0.886 g/cm³ (20 °C)
Flash Pt: 10
Storage Temperature: Ambient
MDL Number: MFCD00009372
CAS Number: 109-60-4
EINECS: 203-686-1
UN: 1276
ADR: 3 II
Merck Index: 12,08026