Ethyl hexanoate, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
148962-100ML 148962-5ML
SIAL148962-100MLEA 47.2 EUR
SIAL148962-100ML SIAL148962-5ML
Ethyl hexanoate, Sigma-Aldrich®
Ethyl hexanoate
Formula: C₈H₁₆O₂
Boiling Pt: 168 °C (1013 hPa)
Melting Pt: –67 °C
Density: 0.87 g/cm³ (20 °C)
Flash Pt: 52 °C
MDL Number: MFCD00009511
CAS Number: 123-66-0
EINECS: 204-640-3
UN: 3272
ADR: 3,III
Merck Index: 13,03812