Ethyl hexanoate, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ300

W243914-1KG-K W243914-4KG-K W243914-9KG-K
SIALW243914-1KG-KEA 135 EUR
SIALW243914-1KG-K SIALW243914-4KG-K SIALW243914-9KG-K
Ethyl hexanoate, Sigma-Aldrich®
Ethyl hexanoate
Formula: C₈H₁₆O₂
Boiling Pt: 168 °C (1013 hPa)
Melting Pt: –67 °C
Density: 0.87 g/cm³ (20 °C)
Flash Pt: 52 °C
MDL Number: MFCD00009511
CAS Number: 123-66-0
EINECS: 204-640-3
UN: 3272
ADR: 3,III
Merck Index: 13,03812

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR