Methyl hexanoate, Sigma-Aldrich®
Supplier: Merck
Synonyms: Caproic acid methyl ester, Methyl caproate, Hexanoic acid methyl ester
M-Clarity™ quality level = MQ300
W270814-100G-K W270814-1KG-K W270814-4KG-K
SIALW270814-100G-KEA 111 EUR
SIALW270814-100G-K SIALW270814-1KG-K SIALW270814-4KG-K
Methyl hexanoate, Sigma-Aldrich®
Methyl hexanoate
Formula: C₇H₁₄O₂
MW: 130.19 g/mol
Boiling Pt: 151 °C (1013 hPa)
Melting Pt: –63 °C
Density: 0.88 g/cm³ (20 °C)
MDL Number: MFCD00009510
CAS Number: 106-70-7
EINECS: 203-425-1
UN: 3272
ADR: 3,III