tert-Butyl formate, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Formic acid tert-butyl ester
M-Clarity™ quality level = MQ200
270776-25G 270776-5G
SIAL270776-25GEA 336 EUR
SIAL270776-25G SIAL270776-5G
tert-Butyl formate, Sigma-Aldrich®
tert-Butyl formate
Formula: C₅H₁₀O₂
MW: 102.13 g/mol
Storage Temperature: Refrigerator
CAS Number: 762-75-4
EINECS: 212-105-0
UN: 3272
ADR: 3 II