Butyl formate, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: Formic acid butyl ester

M-Clarity™ quality level = MQ300

W219606-1KG W219606-5KG W219606-SAMPLE
SIALW219606-1KGEA 136 EUR
SIALW219606-1KG SIALW219606-5KG SIALW219606-SAMPLE
Butyl formate, Sigma-Aldrich®
Butyl formate
Formula: C₅H₁₀O₂
MW: 102.13 g/mol
Boiling Pt: 107
Melting Pt: –92 °C
Density: 0.898 g/cm³ (20 °C)
Flash Pt: 18
Storage Temperature: Ambient
MDL Number: MFCD00003296
CAS Number: 592-84-7
EINECS: 209-772-5
UN: 1128
ADR: 3 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR