Ethyl formate 98+%, pure
Supplier: Thermo Scientific
Danger
Synonyms: Formic acid ethyl ester
150670010 150670025 150675000 150670251
ACRO150670010EA 81.7 EUR
ACRO150670010 ACRO150670025 ACRO150675000 ACRO150670251
Ethyl formate 98+%, pure
Ethyl formate
Formula: C₃H₆O₂
MW: 74.08 g/mol
Boiling Pt: 54 °C (760 mmHg)
Melting Pt: –80 °C
Density: 0.917 g/cm³
Flash Pt: –20 °C
Storage Temperature: Ambient
MDL Number: MFCD00003294
CAS Number: 109-94-4
EINECS: 203-721-0
UN: 1190
ADR: 3,II
Merck Index: 13,03841
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3580 to 1.3630 (20°C, 589 nm)
GC ≥98.0 %