Ethyl formate, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: Formic acid ethyl ester

M-Clarity™ quality level = MQ200

112682-25ML 112682-500ML 112682-2.5L
SIAL112682-25MLEA 43.5 EUR
SIAL112682-25ML SIAL112682-500ML SIAL112682-2.5L
Ethyl formate, Sigma-Aldrich®
Ethyl formate
Formula: C₃H₆O₂
MW: 74.08 g/mol
Boiling Pt: 148 °C (1013 hPa)
Melting Pt: –2 °C
Density: 0.9208 g/cm³ (20 °C)
Flash Pt: –20 °C
Storage Temperature: Ambient
MDL Number: MFCD00003294
CAS Number: 109-94-4
EINECS: 203-721-0
UN: 1190
ADR: 3,II
Merck Index: 13,03841

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR