Ethyl formate, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Formic acid ethyl ester
M-Clarity™ quality level = MQ300
W243418-10KG-K W243418-1KG-K W243418-25KG-K
SIALW243418-10KG-KEA 333 EUR
SIALW243418-10KG-K SIALW243418-1KG-K SIALW243418-25KG-K
Ethyl formate, Sigma-Aldrich®
Ethyl formate
Formula: C₃H₆O₂
Boiling Pt: 148 °C (1013 hPa)
Melting Pt: –2 °C
Density: 0.9208 g/cm³ (20 °C)
Flash Pt: –20 °C
Storage Temperature: Ambient
MDL Number: MFCD00003294
CAS Number: 109-94-4
EINECS: 203-721-0
UN: 1190
ADR: 3,II
Merck Index: 13,03841