Methyl formate 98% for spectroscopy
Supplier: Thermo Fisher Scientific
Danger
Synonyms: Formic acid methyl ester
414345000 414340050 414340025
ACRO414345000EA 84.3 EUR
ACRO414345000 ACRO414340050 ACRO414340025
Methyl formate 98% for spectroscopy
Methyl formate
Formula: C₂H₄O₂
MW: 60.05 g/mol
Boiling Pt: 32
Melting Pt: –100 °C
Density: 0.968 g/cm³
Flash Pt: –32 °C
Storage Temperature: Ambient
MDL Number: MFCD00003291
CAS Number: 107-31-3
EINECS: 203-481-7
UN: 1243
ADR: 3 I
Merck Index: 12,06153
Order Now
Specification Test Results
Appearance Clear liquid
Infrared spectrum Conforms
Water ≤0.05 %
GC ≥97.5 %
UV at 320 nm A: ≤0.009
Color scale ≤10 APHA
Residue after evaporation ≤0.0005 %
Acidity ≤0.0008 meq/g