Ethyl stearate
Supplier: Cayman Chemical
Synonyms: Stearic acid ethyl ester, Ethyl octadecanoate
10008196-100 10008196-500
CAYM10008196-100EA 31 EUR
CAYM10008196-100 CAYM10008196-500
Ethyl stearate
Ethyl stearate
Formula: C₂₀H₄₀O₂
MW: 312.54 g/mol
Boiling Pt: 365
Melting Pt: 31
Density: 0.8973 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00009006
CAS Number: 111-61-5
EINECS: 203-887-4