Ethyl stearate, Sigma-Aldrich®

Supplier: Merck

Synonyms: Stearic acid ethyl ester, Ethyl octadecanoate

M-Clarity™ quality level = MQ300

W349003-1KG W349003-250G W349003-5KG W349003-SAMPLE
SIALW349003-1KGEA 138 EUR
SIALW349003-1KG SIALW349003-250G SIALW349003-5KG SIALW349003-SAMPLE
Ethyl stearate, Sigma-Aldrich®
Ethyl stearate
Formula: C₂₀H₄₀O₂
MW: 312.54 g/mol
Boiling Pt: 365
Melting Pt: 31
Density: 0.8973 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00009006
CAS Number: 111-61-5
EINECS: 203-887-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR