Ethyl octanoate, Sigma-Aldrich®

Supplier: Merck

Synonyms: Caprylic acid ethyl ester, Octanoic acid ethyl ester, Ethyl caprylate

M-Clarity™ quality level = MQ100

112321-100G 112321-5G
SIAL112321-100GEA 28.5 EUR
SIAL112321-100G SIAL112321-5G
Ethyl octanoate, Sigma-Aldrich®
Ethyl octanoate
Formula: C₁₀H₂₀O₂
MW: 172.27 g/mol
Boiling Pt: 206 °C (1013 hPa)
Melting Pt: –47 °C
Density: 0.87 g/cm³ (20 °C)
Flash Pt: 81 °C
Storage Temperature: Ambient
MDL Number: MFCD00009552
CAS Number: 106-32-1
EINECS: 203-385-5
Merck Index: 13,03813

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR