Ethyl octanoate, Sigma-Aldrich®
Supplier: Merck
Synonyms:
Caprylic acid ethyl ester, Octanoic acid ethyl ester, Ethyl caprylate
M-Clarity™ quality level = MQ300
W244902-1KG-K
W244902-20KG-K
W244902-4KG-K
SIALW244902-1KG-KEA
80.3
EUR
SIALW244902-1KG-K
SIALW244902-20KG-K
SIALW244902-4KG-K
Ethyl octanoate, Sigma-Aldrich®
Ethyl octanoate
Formula:
C₁₀H₂₀O₂ MW: 172.27 g/mol Boiling Pt: 206 °C (1013 hPa) Melting Pt: –47 °C Density: 0.87 g/cm³ (20 °C) Flash Pt: 81 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009552 CAS Number: 106-32-1 EINECS: 203-385-5 Merck Index: 13,03813 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...