Methyl octanoate, Sigma-Aldrich®
Supplier: Merck
Synonyms: Caprylic acid methyl ester, Octanoic acid methyl ester, Methyl caprylate
M-Clarity™ quality level = MQ300
W272809-1KG-K W272809-9KG-K
SIALW272809-1KG-KEA 86.3 EUR
SIALW272809-1KG-K SIALW272809-9KG-K
Methyl octanoate, Sigma-Aldrich®
Methyl octanoate
Formula: C₉H₁₈O₂
MW: 158.24 g/mol
Boiling Pt: 190 °C (1013 hPa)
Melting Pt: –36.7 °C
Density: 0.877 g/cm³ (20 °C)
Flash Pt: 68 °C
MDL Number: MFCD00009551
CAS Number: 111-11-5
EINECS: 203-835-0