Butyl propionate 99+%
Supplier: Thermo Scientific
ACRO269482500EA
31.4
EUR
ACRO269482500
ACRO269480010
Butyl propionate 99+%
Butyl propionate
Formula:
C₇H₁₄O₂ MW: 130,19 g/mol Boiling Pt: 145 °C (760 mmHg) Melting Pt: –75 °C Density: 0,88 g/cm³ Flash Pt: 38 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009448 UN: 1914 ADR: 3,III Merck Index: 12,01623 |
Specification Test Results
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4000 to 1.4020 (20°C, 589 nm) |
GC | ≥99.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...