Butyl propionate

Supplier: EHRENSTORFER GMBH

EHERC16493400EA 42.2 EUR
EHERC16493400
Butyl propionate
Butyl propionate
Formula: C₇H₁₄O₂
MW: 130,19 g/mol
Boiling Pt: 145 °C (1013 hPa)
Melting Pt: –90 °C
Density: 0,88 g/cm³ (20 °C)
MDL Number: MFCD00009448
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR