Butyl propionate, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ300
SIALW221104-20KG-KEA 1400 EUR
SIALW221104-20KG-K SIALW221104-9KG-K SIALW221104-1KG-K
Butyl propionate, Sigma-Aldrich®
Butyl propionate
Formula: C₇H₁₄O₂
MW: 130,19 g/mol
Boiling Pt: 145 °C (1013 hPa)
Melting Pt: –90 °C
Density: 0,88 g/cm³ (20 °C)
MDL Number: MFCD00009448
ADR: 3,III
Merck Index: 12,01623