Ethyl propionate, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
Synonyms:
Propionic acid ethyl ester
M-Clarity™ quality level = MQ300
W245607-10KG-K
W245607-1KG-K
W245607-20KG-K
SIALW245607-10KG-KEA
267
EUR
SIALW245607-10KG-K
SIALW245607-1KG-K
SIALW245607-20KG-K
Ethyl propionate, Sigma-Aldrich®
Ethyl propionate
Formula:
C₅H₁₀O₂ Boiling Pt: 210 °C (1013 hPa) Melting Pt: –74 °C Density: 0.89 g/cm³ (20 °C) Flash Pt: 12 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009308 CAS Number: 105-37-3 EINECS: 203-291-4 UN: 1195 ADR: 3,II Merck Index: 13,03878 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...