Ethyl propionate, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Propionic acid ethyl ester
M-Clarity™ quality level = MQ300
W245615-100G-K W245615-1KG-K W245615-5KG-K
SIALW245615-100G-KEA 97.5 EUR
SIALW245615-100G-K SIALW245615-1KG-K SIALW245615-5KG-K
Ethyl propionate, Sigma-Aldrich®
Ethyl propionate
Formula: C₅H₁₀O₂
Boiling Pt: 210 °C (1013 hPa)
Melting Pt: –74 °C
Density: 0.89 g/cm³ (20 °C)
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00009308
CAS Number: 105-37-3
EINECS: 203-291-4
UN: 1195
ADR: 3,II
Merck Index: 13,03878