Methyl propionate 99+%
Supplier: Thermo Scientific
Danger
Synonyms: Propanoic acid methyl ester
149050010 149052500
149050010.EA 125 EUR
149050010. 149052500.
Methyl propionate 99+%
Methyl propionate
Formula: C₄H₈O₂
MW: 88.11 g/mol
Boiling Pt: 79.7 °C (1013 hPa)
Melting Pt: –88 °C
Density: 0.915 g/cm³
Flash Pt: –2 °C
Storage Temperature: Ambient
MDL Number: MFCD00009306
CAS Number: 554-12-1
EINECS: 209-060-4
UN: 1248
ADR: 3,II
Merck Index: 12,06191
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3750 to 1.3790 (20°C, 589 nm)
Water ≤0.3 % (K.F.)
GC ≥99.0 %
Acid value ≤1 mg KOH/g