Isopropyl myristate, Sigma-Aldrich®

Supplier: Merck

Synonyms: Tetradecanoic acid isopropyl ester, Myristic acid isopropyl ester, Isopropyl Myristate, Isopropyl tetradecanoate, Tetradecylic acid isopropyl ester

MQ100

172472-1L 172472-25ML
SIAL172472-1LEA 154 EUR
SIAL172472-1L SIAL172472-25ML
Isopropyl myristate, Sigma-Aldrich®
Isopropyl myristate
Formula: C₁₇H₃₄O₂
MW: 270.46 g/mol
Boiling Pt: 320
Melting Pt: 0…1 °C
Density: 0.852 g/cm³ (20 °C)
Flash Pt: 150
Storage Temperature: Refrigerator
MDL Number: MFCD00008982
CAS Number: 110-27-0
EINECS: 203-751-4
Merck Index: 13,05235

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR