Isopropyl myristate, Sigma-Aldrich®

Supplier: Merck

Synonyms: Tetradecanoic acid isopropyl ester, Myristic acid isopropyl ester, Isopropyl Myristate, Isopropyl tetradecanoate, Tetradecylic acid isopropyl ester

MQ300

W355690-1KG-K W355690-20KG-K W355690-8KG-K
SIALW355690-1KG-KEA 109 EUR
SIALW355690-1KG-K SIALW355690-20KG-K SIALW355690-8KG-K
Isopropyl myristate, Sigma-Aldrich®
Isopropyl myristate
Formula: C₁₇H₃₄O₂
MW: 270.46 g/mol
Boiling Pt: 320
Melting Pt: 0…1 °C
Density: 0.852 g/cm³ (20 °C)
Flash Pt: 150
Storage Temperature: Refrigerator
MDL Number: MFCD00008982
CAS Number: 110-27-0
EINECS: 203-751-4
Merck Index: 13,05235

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR