Methyl myristate, Sigma-Aldrich®

Supplier: Merck

Synonyms: Tetradecylic acid methyl ester, Myristic acid methyl ester, Tetradecanoic acid methyl ester, Methyl tetradecanoate

M-Clarity™ quality level = MQ300

W272205-1KG-K W272205-20KG-K W272205-8KG-K
SIALW272205-1KG-KEA 114 EUR
SIALW272205-1KG-K SIALW272205-20KG-K SIALW272205-8KG-K
Methyl myristate, Sigma-Aldrich®
Methyl myristate
Formula: C₁₅H₃₀O₂
MW: 242.4 g/mol
Boiling Pt: 323
Melting Pt: 18
Density: 0.86715 g/cm³ (20 °C)
Flash Pt: 140
Storage Temperature: Refrigerator
MDL Number: MFCD00008983
CAS Number: 124-10-7
EINECS: 204-680-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR