Dimethyl carbonate, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
SIAL517127-1LEA
166
EUR
SIAL517127-1L
SIAL517127-100ML
SIAL517127-2L
Dimethyl carbonate, Sigma-Aldrich®
Dimethyl carbonate
Formula:
C₃H₆O₃ MW: 90,08 g/mol Boiling Pt: 90 °C (1013 hPa) Melting Pt: 0,5…4.7 °C Density: 1,07 g/cm³ (20 °C) Flash Pt: 18 °C |
UN:
1161 ADR: 3,II Merck Index: 12,06117 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...