Dimethyl carbonate, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ200
D152927-1L D152927-500ML
SIALD152927-1LEA 83.5 EUR
SIALD152927-1L SIALD152927-500ML
Dimethyl carbonate, Sigma-Aldrich®
Dimethyl carbonate
Formula: C₃H₆O₃
MW: 90.08 g/mol
Boiling Pt: 90
Melting Pt: 0.5…4.7 °C
Density: 1.07 g/cm³ (20 °C)
Flash Pt: 18
Storage Temperature: Ambient
MDL Number: MFCD00008420
CAS Number: 616-38-6
EINECS: 210-478-4
UN: 1161
ADR: 3 II
Merck Index: 12,06117