Bis(2-ethylhexyl)sebacate, TECHNICAL
Supplier: VWR Chemicals
23399.296EA 94.2 EUR
23399.296 23399.460
Bis(2-ethylhexyl)sebacate, TECHNICAL
Bis(2-ethylhexyl) sebacate
Formula: C₂₆H₅₀O₄
Boiling Pt: 435 °C (1013 hPa)
Melting Pt: –55 °C
Density: 0.914 g/cm³ (20 °C)
Flash Pt: 213 °C
Storage Temperature: Ambient
MDL Number: MFCD00009497
CAS Number: 122-62-3
EINECS: 204-558-8
Merck Index: 13,01251
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=18b3fddd17e2c2f9148bc413e5e2c4558f3bc0a9&vid=689b9837fe6dd360b0805997dc4becc04db9113e&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Identification Passes test