Dibutyl sebacate, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
Synonyms:
Sebacic acid dibutyl ester, 1,10-Decanedioic acid dibutyl ester, Dibutyl decanedioate
M-Clarity™ quality level = MQ100
D49504-1L
D49504-250ML
SIALD49504-1LEA
160
EUR
SIALD49504-1L
SIALD49504-250ML
Dibutyl sebacate, Sigma-Aldrich®
Dibutyl sebacate
Formula:
C₁₈H₃₄O₄ MW: 314.47 g/mol Boiling Pt: 180 Melting Pt: –11 °C Density: 0.94 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00027218 CAS Number: 109-43-3 EINECS: 203-672-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...