Diethyl sebacate, Sigma-Aldrich®
Supplier: Merck
Synonyms: Sebacic acid diethyl ester, Decanedioic acid diethyl ester, Diethyl decanedioate
M-Clarity™ quality level = MQ300
W237604-10KG-K W237604-1KG-K
SIALW237604-10KG-KEA 1700 EUR
SIALW237604-10KG-K SIALW237604-1KG-K
Diethyl sebacate, Sigma-Aldrich®
Diethyl sebacate
Formula: C₁₄H₂₆O₄
Boiling Pt: 312 °C (1013 hPa)
Melting Pt: –3 °C
Density: 0.9646 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00009219
CAS Number: 110-40-7
EINECS: 203-764-5