Diethyl malonate, Sigma-Aldrich®
Supplier: Merck
Synonyms: 1,3-Propanedioic acid diethyl ester, Malonic acid diethyl ester
M-Clarity™ quality level = MQ200
D97754-1KG D97754-2.5KG D97754-25G D97754-500G
SIALD97754-1KGEA 81.7 EUR
SIALD97754-1KG SIALD97754-2.5KG SIALD97754-25G SIALD97754-500G
Diethyl malonate, Sigma-Aldrich®
Diethyl malonate
Formula: C₇H₁₂O₄
MW: 160.17 g/mol
Boiling Pt: 199
Melting Pt: –50 °C
Density: 1.055 g/cm³ (20 °C)
Flash Pt: 93
Storage Temperature: Ambient
MDL Number: MFCD00009195
CAS Number: 105-53-3
EINECS: 203-305-9
Merck Index: 13,03856