Diethyl succinate, Sigma-Aldrich®
Supplier: Merck
Synonyms: Succinic acid diethyl ester, 1,4-Butanedioic acid diethyl ester
M-Clarity™ quality level = MQ100
112402-1KG 112402-250G
SIAL112402-1KGEA 116 EUR
SIAL112402-1KG SIAL112402-250G
Diethyl succinate, Sigma-Aldrich®
Diethyl succinate
Formula: C₈H₁₄O₄
MW: 174.2 g/mol
Boiling Pt: 217 °C (1013 hPa)
Melting Pt: –19 °C
Density: 1.04 g/cm³ (20 °C)
Flash Pt: 80 °C
Storage Temperature: Ambient
MDL Number: MFCD00009208
CAS Number: 123-25-1
EINECS: 204-612-0
Merck Index: 12,09038