Dimethyl pimelate ≥98%
Supplier: Thermo Scientific
Synonyms: Dimethyl heptanedioate, 1,7-Heptanedioic acid dimethyl ester, Pimelic acid dimethyl ester
B22026.06 B22026.14 B22026.22
B22026.06EA 44.2 EUR
B22026.06 B22026.14 B22026.22
Dimethyl pimelate ≥98%
Dimethyl pimelate
Purity: 99% CAS No.: 1732-08-7 Molecular Formula: C9H16O4 Formula Weight: 188.23 Flash Point: 110°(230°F) Refractive Index: 1.4310 Odour: Mild Boiling Point: 121-122°/11mm Beilstein Reference: 1777309 MDL No.: MFCD00008470
Formula: C₉H₁₆O₄
MW: 188.22 g/mol
Boiling Pt: 121…122 °C (11 mmHg)
Melting Pt: –18.7 °C
Density: 1.041
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008470
CAS Number: 1732-08-7
EINECS: 217-057-4