Diethyl oxalate, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Oxalic acid diethyl ester
M-Clarity™ quality level = MQ100
75712-1L 75712-250ML
SIAL75712-1LEA 82.2 EUR
SIAL75712-1L SIAL75712-250ML
Diethyl oxalate, Sigma-Aldrich®
Diethyl oxalate
Formula: C₆H₁₀O₄
MW: 146.14 g/mol
Boiling Pt: 185.7 °C (1013 hPa)
Melting Pt: –41 °C
Density: 1.0785 g/cm³ (20 °C)
Flash Pt: 76 °C
Storage Temperature: Ambient
MDL Number: MFCD00009119
CAS Number: 95-92-1
EINECS: 202-464-1
UN: 2525
ADR: 6.1,III
Merck Index: 13,03152