1-Naphthyl butyrate, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
N8000-10G N8000-1G N8000-250MG N8000-5G
SIALN8000-10GEA 636 EUR
SIALN8000-10G SIALN8000-1G SIALN8000-250MG SIALN8000-5G
1-Naphthyl butyrate, Sigma-Aldrich®
1-Naphthyl butyrate
Formula: C₁₄H₁₄O₂
Boiling Pt: 125…130 °C (1 mmHg)
Melting Pt: 95.5…96.5 °C
Density: 1.0191 g/cm³ (17.2 °C)
Storage Temperature: Freezer
MDL Number: MFCD00003923
CAS Number: 3121-70-8
EINECS: 221-500-7