1-Naphthyl acetate, Sigma-Aldrich®
Supplier: Merck
Synonyms: Naphthalen-1-yl acetate, Ethanoic acid 1-naphthyl ester, Acetic acid 1-naphthyl ester, α-Naphthyl acetate, Acetic acid α-naphthyl ester, Ethanoic acid α-naphthyl ester
M-Clarity™ quality level = MQ200
N8505-100G N8505-25G N8505-5G
SIALN8505-100GEA 652 EUR
SIALN8505-100G SIALN8505-25G SIALN8505-5G
1-Naphthyl acetate, Sigma-Aldrich®
1-Naphthyl acetate
Formula: C₁₂H₁₀O₂
MW: 186.21 g/mol
Boiling Pt: 310 °C (1013 hPa)
Melting Pt: 41…43 °C
Density: 1.1305 g/cm³ (50 °C)
Flash Pt: >100 °C
Storage Temperature: Freezer
MDL Number: MFCD00003922
CAS Number: 830-81-9
EINECS: 212-599-8