Octanoyl chloride, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ200

O4733-100ML O4733-500ML O4733-5ML
SIALO4733-100MLEA 43.9 EUR
SIALO4733-100ML SIALO4733-500ML SIALO4733-5ML
Octanoyl chloride, Sigma-Aldrich®
Octanoyl chloride
Formula: C₈H₁₅ClO
Boiling Pt: 196 °C (1013 hPa)
Melting Pt: –63 °C
Density: 0.95 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000766
CAS Number: 111-64-8
EINECS: 203-891-6
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR